Learn More
Invitrogen™ SimplyBlue™ SafeStain
SimplyBlue SafeStain is a ready-to-use, fast, sensitive, and safe Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes.
$326.00 - $760.00
Specifications
Content And Storage | SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent. Store at room temperature. Guaranteed stable for 6 months when properly stored. There are no HazMat charges associated with this product. |
---|---|
Detection Location | In-Gel Detection |
Detection Method | Colorimetric |
Label or Dye | Coomassie |
Product Type | Safe Protein Stain |
Includes: SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent.
Description
SimplyBlue SafeStain is a ready-to-use, fast, sensitive, and safe Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes. SimplyBlue SafeStain is completely non-hazardous and does not require methanol or acetic acid fixatives or destains. Risk of hazardous exposure and unpleasant odors are eliminated, creating a healthier lab environment.
Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes. SimplyBlue SafeStain is completely non-hazardous and does not require methanol or acetic acid fixatives or destains. Risk of hazardous exposure and unpleasant odors are eliminated, creating a healthier lab environment.Features include:
• An easy-to-perform protocol that can be completed in less than three hours. For added speed, a simple microwave procedure can be completed in 12 minutes
• Destaining not required but may be performed with deionized water to achieve maximum sensitivity, especially when performing downstream analyses such as mass spectrometry, or when a crystal-clear background is required
• Sensitive—detect down to 7 ng of protein
• Fast—detect 1 μg of protein in as little as five minutes after staining
• Convenient—save valuable time by skipping reagent preparation with this ready-to-use formulation
• Non-hazardous—save money and hassle associated with hazardous shipping and disposal of hazardous waste
Order Info
Shipping Condition: Room Temperature
Specifications
SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent. Store at room temperature. Guaranteed stable for 6 months when properly stored. There are no HazMat charges associated with this product. | |
Colorimetric | |
Safe Protein Stain | |
SimplyBlue™ |
In-Gel Detection | |
Coomassie | |
Less hazardous | |
Protein |