Learn More
FyterTech Nonwovens MERCSORB™ Mercury Amalgamation Powder
Use for dangerous mercury spills
Supplier: FyterTech Nonwovens JV520500
Description
- Amalgamation powder converts elemental mercury into amalgam and stops dangerous mercury vapors from being emitted
- Components: Zinc dust, Citric acid
Specifications
| Zinc Dust, Citric Acid | |
| Amalgamation Powder | |
| Convert elemental mercury into amalgam and stops dangerous mercury vapors from being emitted, for dangerous mercury spills | |
| 500 g |
To be used by trained personnel only