missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nuclease-Free Water, Promega™
Quality tested and certified free of DNase and RNase activity.
Supplier: Promega Corporation P1195
Description
- Quality tested and certified free of DNase and RNase activity
Specifications
Water | |
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 |
Nuclease-Free | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
150 mL |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Safety and Handling
Recommended Storage : Below 30°C