missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nuclease-Free Water, Promega™ DFS Item

Quality tested and certified free of DNase and RNase activity.

Supplier:  Promega Corporation P1199

 View more versions of this product

Catalog No. PRP1199


Explore available promotions

Description

Description

  • Quality tested and certified free of DNase and RNase activity
Specifications

Specifications

Water
7732-18-5
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Nuclease free
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
1000 mL

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Below 30°C

SDS
Documents

Documents

Promotions

Promotions