missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nujol, for IR spectroscopy, Thermo Scientific Chemicals

For IR spectroscopy

Supplier:  Thermo Scientific Chemicals L14480AD

 View more versions of this product

Catalog No. AAL14480AD


Explore available promotions

Description

Description

Nujol is used for the synthetic resin used for cosmetics, fiber oil, food, medicine, as automotive lubricant, home decoration and in candle making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nujol is used for the synthetic resin used for cosmetics, fiber oil, food, medicine, as automotive lubricant, home decoration and in candle making.

Solubility
Soluble in ether. Sparingly soluble in ethanol. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
Specifications

Specifications

Nujol
8012-95-1
0.838
220°C (428°F)
1.468
50 mL
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
FFNDMZIBVDSQFI-UHFFFAOYSA-N
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
68245
IR Spectroscopy
for IR spectroscopy
∼-20°C
>300°C
MFCD00131611
MFCD00131611
14,7023
Soluble in ether. Sparingly soluble in ethanol. Not miscible or difficult to mix in water.
*
0.00
CHEBI:38701

Chemical Identifiers

8012-95-1
0.00
FFNDMZIBVDSQFI-UHFFFAOYSA-N
68245
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
MFCD00131611
MFCD00131611
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
CHEBI:38701
*
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 232-384-2

RTECSNumber : PY8030000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only