Thermo Scientific™ Nunc™ 1536-Well Microplates Plates
Screen four times as many samples in a single plate with Thermo Scientifi™ Nunc™ 1536-Well Microplates, which are ideal for high-throughput screening.
$5052.56 - $5103.41
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid Without Lid
Array 32 x 48
Product Line Nunc
View More Specs
Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
12-565-451
View Documents
Thermo Scientific™
253601
Black
Case of 90 Each for $5,052.56
12-565-452
View Documents
Thermo Scientific™
253607
White
Case of 90 Each for $5,103.41
Description
Description
Maximize high-throughput screening efficiency by using Thermo Scientific™ Nunc™ 1536-Well Microplates, built according to ANSI standards and with the highest quality.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum signal reflection and minimal crosstalk between wells
Warranty: 90 days
Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Without Lid
Nunc
Polystyrene
1 to 10 μL
Non-treated
90
90/Cs.
Standard
32 x 48
Microplate
1536
Non-sterile
Flat Bottom (F-well)
30
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.