missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ 384 Well ShallowWell Standard Height Clear Plates
Reduce costs using Thermo Scientific™ Nunc™ 384-well shallow well standard height clear polystyrene plates—total well volume is reduced to 25μL.
$3454.12 - $3454.12
Specifications
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
---|---|
Format | Standard |
Array | 16 x 24 |
Product Line | Nunc |
Color | Clear |
Catalog Number | Mfr. No. | Volume (Metric) Well | No. per Pack | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) Well | No. per Pack | Price | Quantity | ||||
12-565-745
|
Thermo Scientific™
264704 |
25 μL | 25 |
Case of 100 Each for $3,454.12
|
|
||||
Description
Accommodate small sample volumes by using Thermo Scientific™ Nunc™ 384-Well ShallowWell Standard Height Clear Plates with a total well volume of 25μL, ideal for high throughput screenings. The standard plate height allows for easy adaptation to most automation systems.
Features:- Non-treated hydrophobic polystyrene plates
- ANSI™ (SBS) standard footprint
- Suitable for colorimetric, fluorescent and luminescent assays
- Customized bar coding available
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
16 x 24 | |
Clear | |
Polystyrene | |
384 | |
100 |
Standard | |
Nunc | |
Microplate | |
25 μL | |
2 to 20 μL | |
25 |