Thermo Scientific™ Nunc™ 96-Well, Nunclon Delta-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Supplier:  Thermo Scientific™ 168136
Catalog No. 12-565-720
Explore available promotions
Description
Description
Thermo Scientific™ Nunc 96-Well Polystyrene Round Bottom Microwell Plates are optimized for robotics, automated readers and liquid handling systems. 96-well round bottom (U) microplates are ideal for agglutination assays.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Specifications
Custom Barcoding Available
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
8 x 12
Nunc
Microplate
300 μL
Sterile
Round Bottom (U-well)
10
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
With Lid
10 Pk., 160 Case
Clear
Polystyrene
96
Nunclon Delta
160
Product Suggestions
Product Suggestions