Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
$925.76 - $3147.24
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Product Line | Nunc |
Product Type | Microplate |
Material | Polystyrene |
Catalog Number | Mfr. No. | Lid | Sterility | No. per Pack | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lid | Sterility | No. per Pack | Price | Quantity | |||
12-565-212
|
Thermo Scientific™
262162 |
Without Lid | Sterile | 1 |
Case of 50 Each for $925.76
|
|
|||
12-565-213
|
Thermo Scientific™
262146 |
Without Lid | Non-sterile | 135 |
Case of 135 Each for $1,655.47
|
|
|||
12-565-214
|
Thermo Scientific™
268152 |
Without Lid | Non-sterile | 10 |
Case of 180 Each for $2,229.09
|
|
|||
12-565-331
|
Thermo Scientific™
268200 |
With Lid | Sterile | 10 |
Case of 160 Each for $3,147.24
|
|
|||
Description
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
- Clear polystyrene has excellent optical properties ideal for optical measurements
- Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
- Customized bar coding is available
Warranty: 90 days
Specifications
Standard | |
Nunc | |
Polystyrene | |
Non-treated |
8 x 12 | |
Microplate | |
96 | |
Round Bottom (U-well) |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.