Thermo Scientific™ Nunc™ Barcode Scanners, USB
Track samples simply using these bar code scanners. Plug and Play function can automatically switch to read codes: 128A, 128B, 128C, 39 and Interleve 2/5.
Supplier: Thermo Scientific™ 330050
Description
Track samples simply using Thermo Scientific™ Nunc™ Barcode Scanners. “Plug and Play” function can automatically switch to read codes: 128A, 128B, 128C, 39 and Interleve 2/5. Convenient and easy to use with an Excel spread sheet.
Durable, low cost, and easy to use–simply plug into a USB or keyboard port.- Scanners automatically switch between Code 128, Code 39, Interleave 2 of 5 and several other barcodes
- Must be in contact with the barcode to read