Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps
Achieve maximum adhesion for a broad range of cell types.
$907.94 - $1242.76
Specifications
Material Polystyrene
Sterility Sterile
View More Specs
Products 2
Catalog Number Mfr. No. Culture Area Closure Type Price Quantity  
Catalog Number Mfr. No. Culture Area Closure Type Price Quantity  
12-565-50
View Documents
Thermo Scientific™
153732
80 cm² Solid
Case of 50 Each for $907.94
12-565-54
View Documents
Thermo Scientific™
156502
175 cm² Solid
Case of 32 Each for $1,242.76
Description
Description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. The vent/close caps with Y marks allow easy visual verification of venting position while promoting sufficient gas exchange.
Features:
  • Nunclon Delta Certification including testing on four different cell types is provided in each case.
  • Short wide necks, straight side walls and 1/3-turn caps are optimized for robotic operation.
  • Vent/close caps with “Y” marks - any of the legs in the “Y” pointing up indicates cap in venting position.
  • Sterilized by gamma irradiation and certified to be non-pyrogenic.
Warranty: 90 days
Specifications
Specifications
Polystyrene
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.