missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks

Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.

Supplier:  Thermo Scientific™ 132867

 View more versions of this product

Catalog No. 12-565-24


Explore available promotions

Description

Description

Perform cell culture expansion and experience 500cm2 total growth area within the footprint of a standard flask using Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.

Three layers of growth surfaces provide a total culture area of 500 cm2 in the same footprint as a standard 175cm2 flask.

  • Recommended working volume is 200mL
  • Promotes conventional, flat, monolayer culturing
  • External dimensions of 175cm2
  • Also offered barcoded with large Code 128

Includes:

Nunclon Delta Certification including testing on four different cell lines is included in each case.

Specifications

Specifications

No
Straight
1
Sterile
Solid
Cell Culture-treated
Cell Culture Treated TripleFlasks
32
Same external dimensions as the standard Nunc T175 with straight neck
Flat
500 cm²
200 mL
Videos
SDS
Documents

Documents

Promotions

Promotions

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.