Thermo Scientific™ Nunc™ Lab-Tek™ Chambered Coverglass
Perform high-magnification microscopy and confocal image analysis with Thermo Scientific™ Nunc™ Lab-Tek™ Chambered Coverglass.
$592.76 - $743.21
Specifications
Lid Material Polystyrene
Surface Treatment Cell Culture
Material Borosilicate Glass (1.0)
Thickness (Metric) 0.13 to 0.17 mm
Includes Clear polystyrene lid with handle on the side for aseptic manipulations
View More Specs
Products 4
Catalog Number Mfr. No. Wells Material Description Price Quantity  
Catalog Number Mfr. No. Wells Material Description Price Quantity  
12-565-472
View Documents
Thermo Scientific™
155361
1 Borosilicate Glass (1.0) 1-well Chambered Coverglass w/ non-removable well
Pack of 16 Each for $743.21
Case of 6 Pack for $4,459.26
12-565-471
View Documents
Thermo Scientific™
155380
2 Borosilicate Glass (1.0) 2-well Chambered Coverglass w/ non-removable wells
Pack of 16 Each for $592.76
Case of 6 Pack for $3,556.56
12-565-401
View Documents
Thermo Scientific™
155383
4 Borosilicate Glass (1.0) 4-well Chambered Coverglass w/ non-removable wells
Pack of 16 Each for $629.59
Case of 6 Pack for $3,777.54
12-565-470
View Documents
Thermo Scientific™
155411
8 Borosilicate Glass (1.0) 8-well Chambered Coverglass w/ non-removable wells
Pack of 16 Each for $633.26
Case of 6 Pack for $3,799.56
Description
Description
Grow, stain and view living cells on a single device. Thermo Scientific™ Nunc™ Lab-Tek™ Chambered Coverglass with a No. 1 borosilicate glass bottom offers the optical characteristics required for high magnification and confocal image analysis. Available in 1-, 2-, 4-, and 8-well formats. The upper chamber structure cannot be removed from the coverglass.
Features:
  • Media chamber securely mounted to No. 1 borosilicate coverglass by non-toxic medical grade silicone adhesive
  • The polystyrene chamber is not removable and requires inverted microscope for viewing
  • Fitted polystyrene cover without top handle minimizes evaporation
Ordering Information:
  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
Specifications
Specifications
Polystyrene
Borosilicate Glass (1.0)
Clear polystyrene lid with handle on the side for aseptic manipulations
57 x 25 mm
Clear
Cell Culture
0.13 to 0.17 mm
16
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.