Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass
Culture and view living cells on a single device. This coverglass has optimum optical characteristics for high-magnification microscopy and confocal image analysis.
$726.29 - $840.21
Specifications
Lid Material Polystyrene
Surface Treatment Cell Culture
Material Borosilicate Glass (1.5)
Thickness (Metric) 0.16 to 0.19 mm
Includes Clear polystyrene lid with handles on the top and side for aseptic manipulations
View More Specs
Products 4
Catalog Number Mfr. No. Wells Material Description Price Quantity  
Catalog Number Mfr. No. Wells Material Description Price Quantity  
12-565-335
View Documents
Thermo Scientific™
155360
1 Borosilicate Glass (1.5) 1-well Chambered Coverglass w/ non-removable well
Pack of 16 Each for $840.21
Case of 6 Pack for $5,041.26
12-565-336
View Documents
Thermo Scientific™
155379
2 Borosilicate Glass (1.5) 2-well Chambered Coverglass w/ non-removable wells
Pack of 16 Each for $817.62
Case of 6 Pack for $4,905.72
12-565-337
View Documents
Thermo Scientific™
155382
4 Borosilicate Glass (1.5) 4-well Chambered Coverglass w/ non-removable wells
Pack of 16 Each for $726.29
Case of 6 Pack for $4,357.74
12-565-338
View Documents
Thermo Scientific™
155409
8 Borosilicate Glass (1.5) 8-well Chambered Coverglass w/ non-removable wells
Pack of 16 Each for $761.94
Case of 6 Pack for $4,571.64
Description
Description
Culture, examine, and analyze living cells on a single device. Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass with a No. 1.5 borosilicate glass bottom offers the ideal optical characteristics required for high-magnification microscopy and confocal image analysis. Available in 1-, 2-, 4-, and 8-well formats. The upper chamber structure cannot be removed from the coverglass.
Features:
  • Media chamber securely mounted to the No. 1.5 borosilicate coverglass by biocompatible acrylic adhesive
  • The polystyrene chamber is not removable and requires inverted microscope for viewing
  • Gas exchange achieved with minimal evaporation through tabbed polystyrene cover
Ordering Information:
  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
Specifications
Specifications
Polystyrene
Borosilicate Glass (1.5)
Clear polystyrene lid with handles on the top and side for aseptic manipulations
55 x 24 mm
Clear
Cell Culture
0.16 to 0.19 mm
16
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.