Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
Supplier:  Thermo Scientific™ 168055
 View more versions of this product
Catalog No. 12-565-64
Only null left
Explore available promotions
Description
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Specifications
Custom Barcoding Available
Without Lid
8 x 12
Nunc
Microplate
Polystyrene
96
Nunclon Delta
Flat Bottom (F-well)
1 Microplate
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
96F well plate, Nunclon Delta, clear, without lid, Sterile (Pack of 1, Case of 50)
1/Pk., 50/Cs.
Clear
High Flange
400 μL
Sterile
400 μL
50 Microplates
50/Cs.
Product Suggestions
Product Suggestions