Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: White; Sterility: Sterile; Lid: With Lid
Supplier:  Thermo Scientific™ 136102
 View more versions of this product
Catalog No. 12-566-26
Only null left
Explore available promotions
Description
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ White Polystyrene Plate. White plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics.
  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
Specifications
Specifications
Custom Barcoding Available
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
8 x 12
Nunc
Microplate
Polystyrene
96
Sterile
Flat Bottom (F-well)
10 Microplates
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
With Lid
10 Pk., 160 Cases
White
Low Flange Pinchbar
400 μL
50 to 200 μL
Nunclon Delta
160 Microplates
Product Suggestions
Product Suggestions