Thermo Scientific™ 96 Well Black Plate, Coverglass Bottom, CC2 Surface, Pack of 6
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Coverglass Base
Supplier:  Thermo Scientific™ 160376
Catalog No. 12-565-607
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available
Specifications
Specifications
Custom Barcoding Available
With Lid
6/Pk., 30/Cs.
Black
High Flange
400 μL
Sterile
Flat Bottom (F-well)
6
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
8 x 12
Nunc
Optical Coverglass Base
Polystyrene
96
CC2, Poly-D Lysine Mimic-Treated
30
Case of 30