Thermo Scientific™ 96 Well Black/Clear Bottom Plate, Poly-D-Lysine Coated Surface, Pack of 5
Color:Black, Clear Bottom; Lid:With Lid; Type: Optical Polymer Base
Supplier:  Thermo Scientific™ 152037
Catalog No. 12-565-902
Explore available promotions
Description
Description
Thermo Scientific™ Nunc MicroWell 96-Well Optical-Bottom Plates with Polymer Base have a solid polystyrene black or white upper structures with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
White or black polystyrene upper structure with polymer or coverglass bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Available in black for fluorescence assays or white for luminescence and fluorescence assays; both with clear bottoms
  • Polymer base plate
  • Working range, 50-200μL/well
  • Customized bar coding is available
  • Aseptically Manufactured
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.
Specifications
Specifications
Polystyrene
50 to 200 μL
Case of 20
20
Poly-D-Lysine
96
8 x 12
5/Pack, 20/Case
Flat
400 μL
Optical Polymer Base
5
Black
Aseptically Manufactured
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Nunc™
Custom Barcoding Available
Product Suggestions
Product Suggestions