Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Supplier:  Thermo Scientific™ 438733
 View more versions of this product
Catalog No. 12-565-154
Explore available promotions
Description
Description
Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
  • Limited adsorption of proteins
  • Lids have keyed tabs for proper orientation
  • Large raised alphanumeric coordinates
  • Unique surface treatment to maximize sample and reagent mixing
  • MiniTrays for serological applications have slightly smaller well dimensions
Specifications
Specifications
Closure Included
MicroWell MiniTray for serological applications
Polystyrene
Non-sterile
Cell Culture Dish
For serological applications
100
84 mm
59 mm
Polystyrene
Flat
Rectangular
Cell Culture
72
Lid
10
59 mm