missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Supplier: Thermo Scientific™ 452256
Description
Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
- Limited adsorption of proteins
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximize sample and reagent mixing
- MiniTrays for serological applications have slightly smaller well dimensions
Specifications
Closure Included | |
MicroWell MiniTray | |
Polystyrene | |
Sterile | |
Cell Culture Dish | |
Lid | |
120 | |
59 mm |
Polystyrene | |
Flat | |
Rectangular | |
Nunclon Delta | |
60 | |
480 | |
84 mm | |
59 mm |