Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems
Take traditional cryostorage to the next level, with standard microplate format racks, accommodating denser storage capability compared to standard cryoboxes.
$597.70 - $3438.80
Specifications
Coding None
Vial Type Cryobank
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Closure Color Sterility Packaging Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Color Sterility Packaging Price Quantity  
12-570-201
View Documents
Thermo Scientific™
374083
0.5 mL Clear Sterile Racked, 96 x 2
Case of 192 Each for $597.70
12-570-209
View Documents
Thermo Scientific™
374087
0.5 mL Clear Sterile Racked, 96 x 10
Case of 960 Each for $2,956.30
12-570-213
View Documents
Thermo Scientific™
374089
1 mL Clear Sterile Racked, 96 x 10
Case of 960 Each for $3,001.52
12-565-282
View Documents
Thermo Scientific™
374222
5 mL Clear Sterile Racked, 48
Case of 480 Each for $2,708.09
12-565-283
View Documents
Thermo Scientific™
374223
2 mL Clear Sterile Racked, 96 x 2
Case of 480 Each for $2,665.06
12-570-228
View Documents
Thermo Scientific™
374260
5 mL Clear Sterile Bulk, 11 bags of 48 tubes/case
Case of 528 Each for $3,438.80
Description
Description
Store diagnostic and biological samples down to vapor-phase liquid nitrogen in Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems, compatible with capping/decapping equipment and labeling options. Cryobank products are available in a volume range of 0.5L through 5.0L, and complimented by a range of certifications.
Secure Tracking
  • Solid one-piece construction eliminates the risk of labels falling off during cryogenic storage, preserving the identity of valuable sample material
Superior Storage Format
  • Cryobank tubes are available in space-conscious microplate format racks or can be stored in traditional cryoboxes, accommodating your preferred storage format
  • Microplate format is compatible with your existing multichannel, manual pipettes and automated liquid-handling equipment
  • Unique anti-rotation feature allows compatibility with automated decapping/capping equipment in single, 8-channel, or full-rack options to accommodate multiple throughput requirements
  • TPE-gasketed polypropylene cap provides superior seal during high- or low-temperature storage
  • Low-binding surface allows maximum recovery of low-concentration samples
Certified to meet your storage needs
  • CE certified and conforms to IATA requirements for diagnostic use and transportation
  • Certified DNAse/RNAse free (0.5/1.0mL), non-pyrongenic (LAL-test), non-toxic (USP class VI test)
  • Sterility assurance level 10-6 for the most sensitive sample storage
Stringent Quality Control
  • Leak-testing validation performed during the manufacturing process to ensure sample integrity is maintained
Specifications
Specifications
None
Cryobank
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'