missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates

Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.

$810.32 - $1549.26

Specifications

Material Polycarbonate (Membrane)
Type Culture Insert
Color Clear
Surface Treatment Cell culture-treated
Sterility Sterile
View More Specs

Products 9
Catalog Number Mfr. No. Pore Size Dish Type Culture Area Price Quantity  
Catalog Number Mfr. No. Pore Size Dish Type Culture Area Price Quantity  
12565006
View Documents
Thermo Scientific™
140620
0.4 μm 24-Well Plate 0.47 cm2
Case of 48 Each for $1,549.26
 
12-565-007
View Documents
Thermo Scientific™
140627
3 μm 24-Well Plate 0.47 cm2
Case of 48 Each for $1,436.32
 
12565008
View Documents
Thermo Scientific™
140629
8 μm 24-Well Plate 0.47 cm2
Case of 48 Each for $1,461.74
 
12-565-012
View Documents
Thermo Scientific™
140640
0.4 μm 6-Well Plate 3.14 cm2
Case of 24 Each for $810.32
 
12-565-013
View Documents
Thermo Scientific™
140642
3 μm 6-Well Plate 3.14 cm2
Case of 24 Each for $842.09
 
12-565-009
View Documents
Thermo Scientific™
140652
0.4 μm 12-Well Plate 1.13 cm2
Case of 48 Each for $1,277.91
 
12-565-010
View Documents
Thermo Scientific™
140654
3 μm 12-Well Plate 1.13 cm2
Case of 48 Each for $1,534.82
 
12565015
View Documents
Thermo Scientific™
140660
0.4 μm 6-Well Plate 4.1 cm2
Case of 24 Each for $853.26
 
12-565-017
View Documents
Thermo Scientific™
140668
8 μm 6-Well Plate 4.1 cm2
Case of 24 Each for $819.85
 
Description

Description

Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.

Features:
  • Tissue culture-treated porous membrane for optimized cell attachment
  • Three pore sizes: 0.4, 3, and 8 micron
  • Pre-packed in sterile Nunclon™ Delta-treated multi-well plates
  • Nontoxic and nonpyrogenic

Applications:

  • Tumor migration and invasion assays
  • Epithelial (e.g. Caco-2) and endothelial (e.g. brain microvascular endothelial cells) barriers assays
  • Dermal/epidermal and epithelial tissue models (e.g. artificial skin model)
  • Chemotaxis studies
  • Co-culture

Warranty: 90 days

 

Specifications

Specifications

Polycarbonate (Membrane)
Clear
Sterile
Culture Insert
Cell culture-treated
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.