missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Supplier: Thermo Scientific™ 236272
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
White | |
Sealng Tape Wht Vinyl Acrylic Adhes | |
Non-sterile | |
96 | |
50 | |
General | |
Nunc | |
No | |
-40°C to +80°C | |
All Microplates | |
Yes |
Life Science Reagent Resistant | |
Vinyl | |
Not Autoclavable | |
50 | |
Luminescence Assays, Light reflective, ELISPOT assays, designed for either top or bottom of optical plates | |
Adhesive Seal | |
Acrylic | |
No | |
All well shapes | |
General |