Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Supplier:  Thermo Scientific™ 232698
 View more versions of this product
Catalog No. 12-565-398
Explore available promotions
Description
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Specifications
Foil
AL SEAL TAPE FOR 96 WELL PLT
Non-sterile
96
100
PCR and storage in 96 well plates
Adhesive Seal
Acrylic
Yes
All well shapes
General
Life Science Reagent Resistant
Aluminum
Not Autoclavable
100
Malleable allows for visual verification of seal security, minimizes evaporation and cross contamination
General; Long term
Nunc
Yes
-80°C to +120°C
All Microplates
Yes