Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Supplier:  Thermo Scientific™ 276014
 View more versions of this product
Catalog No. 12-565-475
Explore available promotions
Description
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Specifications
Foil
ALUMINUM SEALING TAPE
Non-sterile
96
100
General; Long term
Nunc
Yes
-80°C to +120°C
All Microplates
Yes
Life Science Reagents Resistant, DMSO Resistant
Aluminum
Not Autoclavable
100
PCR; Excellent chemical resistance, Minimal sample evapration, Malleable allows for visual verification of seal security
Adhesive Seal
Acrylic
Yes
All well shapes
General
SDS
Documents
Documents
Accessories