Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Supplier:  Thermo Scientific™ 235307
 View more versions of this product
Catalog No. 12-565-513
Explore available promotions
Description
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Specifications
Clear
SEALING TAPE ADV POLYOLEFIN
Non-sterile
96
100
General; Long term
Nunc
Yes
-80°C to +105°C
All Microplates
Yes
Life Science Reagents Resistant, DMSO Resistant
Polyolefin
Not Autoclavable
100
PCR; qPCR; ELISA; Protein crystallography
Adhesive Seal
Silicone, Non-tacky adhesive, Strong adhesive bond
No
All well shapes
General
Low Autofluorescence
SDS
Documents
Documents
Accessories