Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Supplier:  Thermo Scientific™ 241205
 View more versions of this product
Catalog No. 12-567-05
Explore available promotions
Description
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
Specifications
White
Sealing Tape Rayon White Ster
Sterile
96
25
Adhesive Seal
Acrylic
Yes
All well shapes
General
Yes
Life Science Reagent Resistant
Non-woven Rayon
Not Autoclavable
50
Cell growth, Bacterial growth, Non cytotoxic, Moisture vapor transmission rate of 4.200 g/m^2/24 hrs, highly gas permeable
Nunc
No
-20°C to +80°C
All Microplates
Yes