Learn More
Thermo Scientific™ Nunc™ Storage Box and Rack
Hold up to 100 vials from 0.5 to 2.0mL in size in the Thermo Scientific™ Nunc™ Storage Box, or 24 vials in a 4x6 array in the Nunc Storage Racks.
Supplier: Thermo Scientific™ 264263
Alert:
Do not use the vials for storage in the liquid phase of liquid nitrogen. Such use may cause liquified nitrogen to be trapped inside the vial and lead to pressure build-up resulting in possible explosion or bio-hazard.
Description
The Thermo Scientific™ Nunc™ Storage Box holds up to 100 Thermo Scientific Nunc Storage Vials from 0.5 to 2.0mL in size (Cat# 264262 and 264300). The polycarbonate storage box is keyed for one way on/off fit of the lid that prevents orientation errors. The see-through top makes it easy to view contents. Boxes can be stacked for compact storage. Autoclavable.
Storage Vial Box
- White polycarbonate with clear lid makes it easy to see contents
- 10×10 array
- L × W × H: 5.7 × 5.7 × 2.4″ (144 × 144 × 62mm)
Specifications
Vial storage box | |
Polycarbonate | |
Cryo Storage Box | |
6/Pk., 12/Cs. | |
5.7 x 5.7 x 2.44 in. (144 x 144 x 62 mm) | |
144 mm | |
144 mm | |
62 mm |
White/Clear Lid | |
Square | |
100 Vials | |
10 x 10 | |
5.7 in. | |
5.7 in. | |
2.44 in. | |
4.4 kg |
The PCR (polymerase chain reaction) process is covered by US patents owned by Hoffmann-La Roche Ltd.