missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Storage Vials with Caps

Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.

$934.02 - $987.66

Specifications

Material PPCO
Sterility Sterile
No. per Case 500
No. per Pack 500
Product Line Nunc
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Working Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Working Price Quantity  
09-740-71A
View Documents
Thermo Scientific™
264262
0.5 mL
Case of 500 Each for $987.66
Only null left
 
0974071B
View Documents
Thermo Scientific™
264300
2.0 mL
Case of 500 Each for $934.02
Only null left
 
Description

Description

Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.

Features:

  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C
  • RNase/DNase-free

Ordering Information:

Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.

Specifications

Specifications

PPCO
500
Nunc
Screw Cap
Certificate of Analysis (COA), Certificate of Conformance (COC)
Sterile
500
Polyethylene
500 Cs.
Storage Vial with Cap
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'