missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AlphaProTech Critical Cover™ NuTech™ Sleeves

Microporous sleeves are designed with sonic welded seams for added durability and protection.

$192.42 - $219.64

Specifications

Construction Tapered
Product Type Sleeve Protector
Material Microporous Laminate
Color White
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-810-577
View Documents
AlphaProTech
MS016064
X-Large
Case of 300 Each for $219.64
 
19-810-578
View Documents
AlphaProTech
MS016068
Universal
Case of 300 Each for $192.42
 
Description

Description

  • Designed with sonic welded seams for added durability and protection
  • For use in many general-use applications
  • Tapered design with elastic at both ends
Specifications

Specifications

Tapered
Microporous Laminate
Non-sterile
Elastic Ends
Elastic
MS-01606
21C CFR Part 820
300
Sleeve Protector
White
Single-use
Ultrasonically Welded
Yes
Cleanrooms/Controlled Environments
Bulk
Class 10,000 (ISO 7)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'