Thermo Scientific™ Nuts for Capillary GC Columns Connections
Use Thermo Scientific™ nuts with compatible ferrules to eliminate leaks and connect your capillary GC columns to your instrument.
$159.98 - $159.98
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
03-050-223
|
Thermo Scientific™
290BT239 |
Brass Nut For 15% Graphite/85% Vespel Ferrules | Graphite/Vespel Ferrules |
Pack of 2 Each for $159.98
|
|
||||
Description
Use Thermo Scientific™ nuts with compatible ferrules to eliminate leaks and connect your capillary GC columns to your instrument. Thermo Scientific™ ferrules are available in three different materials - 100% Graphite, 15% Graphite / 85% Vespel, or SilTite Metal - and various dimensions to match your instrument and capillary column inner diameter. Use the nut that is compatible with your ferrule.
Specifications
Capillary Column Nut |