missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nystatin, Thermo Scientific Chemicals

Nystatin, CAS # 1400-61-9, is a polyene compound that shows anti-bacterial and antifungal activities. It is also an ionophore that increases the membrane permeability to some ions.

$141.70 - $246.53

Chemical Identifiers

CAS 1400-61-9
Molecular Formula C47H75NO17
Molecular Weight (g/mol) 926.107
MDL Number MFCD00036240
InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
PubChem CID 133640190
IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6248606
View Documents
Thermo Scientific Chemicals
J6248606
5 g
Each for $141.70
 
AAJ6248609
View Documents
Thermo Scientific Chemicals
J6248609
10 g
Each for $246.53
 
Description

Description

Nystatin which acts a polyene antifungal and antibiotic which increases membrane permeability. Nystatin is used to treat fungal infections of the skin, mouth, vagina, and intestinal tract. Fungal medicines will not work for colds, flu, or other viral infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Nystatin is a polyene compound that shows anti-bacterial and antifungal activities against Streptomyces noursei, Streptomyces aureus, and other Streptomyces species
  • It is also an ionophore with the capacity to increase membrane permeability
  • Nystatin is a channel-forming ionophore and exerts its activity via formation of a membrane-spanning pore in the fungal plasma membrane, thus altering membrane permeability, allowing the leakage of intracellular contents, and the subsequent disruption of electrochemical gradients essential for proper cell function

Applications

  • In in vitro experiments, Nystatin is an antifungal that is both fungistatic and fungicidal against a wide variety of yeasts and yeast-like fungi
  • It has a higher selectivity for fungal cells over mammalian cells since nystatin presents greater binding affinity with ergosterol, a key sterol found in fungal cell walls, as opposed to its mammalian counterpart, cholesterol
  • This compound is not active against organisms that do not contain sterols in their cell membrane (for example, bacteria)
  • It is used on human macrophage cell line U937, to study its effect on the production of interleukin-8 by silver nanoparticle-treated U937 cell line
  • It has the ability to prevent cell culture contamination as well as induce interleukin-1 and -8, and tumor necrosis factor α secretion in TLR2-expressing THP1 cells
  • It modulates lipid raft profile in mice macrophages
Specifications

Chemical Identifiers

1400-61-9
926.107
VQOXZBDYSJBXMA-AWCIUYGFSA-N
(1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
C47H75NO17
MFCD00036240
133640190
CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O

Specifications

1400-61-9
White to Yellow
MFCD00036240
14,6736
VQOXZBDYSJBXMA-AWCIUYGFSA-N
(1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
133640190
Fine Powder
160°C
C47H75NO17
5 g
Soluble in DMSO
CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
926.107
926.09
Nystatin
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-749-0

RTECSNumber : RF5950000

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only