missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene, Thermo Scientific Chemicals

$257.74 - $990.11

Chemical Identifiers

CAS 136157-98-7
Molecular Formula C50H64O8
Molecular Weight (g/mol) 793.054
MDL Number MFCD00798565
InChI Key GRSQCGICFHAMOA-UHFFFAOYSA-N
Synonym o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene, o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-t-butylcalix 4 arene
PubChem CID 15243456
SMILES CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3OCC(=O)O)CC4=CC(=CC(=C4OC)CC5=C(C(=CC(=C5)C(C)(C)C)C2)OCC(=O)O)C(C)(C)C)C(C)(C)C)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1919277
View Documents
Thermo Scientific Chemicals
A1919277
0.1 g N/A
 
AAA1919202
View Documents
Thermo Scientific Chemicals
A1919202
0.5 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

136157-98-7
793.054
GRSQCGICFHAMOA-UHFFFAOYSA-N
15243456
C50H64O8
MFCD00798565
o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene, o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-t-butylcalix 4 arene
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3OCC(=O)O)CC4=CC(=CC(=C4OC)CC5=C(C(=CC(=C5)C(C)(C)C)C2)OCC(=O)O)C(C)(C)C)C(C)(C)C)OC

Specifications

136157-98-7
C50H64O8
0.1 g
GRSQCGICFHAMOA-UHFFFAOYSA-N
793.054
793.06
250°C to 252°C
MFCD00798565
o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene, o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-t-butylcalix 4 arene
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3OCC(=O)O)CC4=CC(=CC(=C4OC)CC5=C(C(=CC(=C5)C(C)(C)C)C2)OCC(=O)O)C(C)(C)C)C(C)(C)C)OC
15243456
O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only