Learn More
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9
Supplier: Thermo Scientific Chemicals 186710050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride | |
227.0°C | |
97.5 to 102.5% (Argentometry) | |
C6F5CH2ONH2·HCl | |
5 g | |
Solubility in water: soluble. | |
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F | |
249.57 | |
249.57 |
57981-02-9 | |
Authentic | |
C7H5ClF5NO | |
MFCD00012953 | |
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv | |
HVMVKNXIMUCYJA-UHFFFAOYSA-N | |
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride | |
122307 | |
98% |
Chemical Identifiers
57981-02-9 | |
249.57 | |
HVMVKNXIMUCYJA-UHFFFAOYSA-N | |
122307 | |
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F |
C7H5ClF5NO | |
MFCD00012953 | |
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv | |
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
EINECSNumber : 261-057-7
RUO – Research Use Only