missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

$123.10 - $438.29

Chemical Identifiers

CAS 876-33-5
MDL Number MFCD01114582
Synonym o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
PubChem CID 120426
IUPAC Name O-[(4-methoxyphenyl)methyl]hydroxylamine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6331203
View Documents
Thermo Scientific Chemicals
H6331203
1 g
Each for $123.10
 
AAH6331206
View Documents
Thermo Scientific Chemicals
H6331206
5 g
Each for $438.29
 
Description

Description

O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

876-33-5
o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
O-[(4-methoxyphenyl)methyl]hydroxylamine
MFCD01114582
120426

Specifications

876-33-5
C8H11NO2
1 g
MVSMBIBGGPSEHQ-UHFFFAOYSA-N
O-[(4-methoxyphenyl)methyl]hydroxylamine
120426
98%
O-(4-Methoxybenzyl)hydroxylamine hydrochloride
White
MFCD01114582
o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
COC1=CC=C(C=C1)CON
153.181
153.18
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

RTECSNumber : NC3676000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only