missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-4-Nitrobenzylhydroxylamine Hydrochloride 99.0+%, TCI America™
Supplier: TCI America A55325G
Specifications
O-4-Nitrobenzylhydroxylamine Hydrochloride [for HPLC Labeling] | |
210°C | |
C7H9ClN2O3 | |
5 g | |
LKCAFSOYOMFQSL-UHFFFAOYSA-N | |
hydrogen O-[(4-nitrophenyl)methyl]hydroxylamine chloride | |
74967 | |
≥99.0% (HPLC) |
2086-26-2 | |
White-Yellow | |
MFCD00012954 | |
o-4-nitrobenzyl hydroxylamine hydrochloride, 4-nitrobenzyloxyamine, o-4-nitrophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-4-nitrophenyl methyl-, monohydrochloride, p-nitrobenzyloxyamine hydrochloride, hydroxylamine, o-p-nitrobenzyl-, monohydrochloride, o-p-nitrobenzyl hydroxylamine hydrochloride, o-4-nitrobenzyl hydroxylammonium hydrochloride, 1-aminooxy methyl-4-nitrobenzene hydrochloride, hydroxylamine, o-p-nitrophenylmethyl-, monohydrochloride | |
[H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O | |
204.61 | |
204.61 | |
Crystalline Powder |
Chemical Identifiers
2086-26-2 | |
204.61 | |
LKCAFSOYOMFQSL-UHFFFAOYSA-N | |
74967 | |
[H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O |
C7H9ClN2O3 | |
MFCD00012954 | |
o-4-nitrobenzyl hydroxylamine hydrochloride, 4-nitrobenzyloxyamine, o-4-nitrophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-4-nitrophenyl methyl-, monohydrochloride, p-nitrobenzyloxyamine hydrochloride, hydroxylamine, o-p-nitrobenzyl-, monohydrochloride, o-p-nitrobenzyl hydroxylamine hydrochloride, o-4-nitrobenzyl hydroxylammonium hydrochloride, 1-aminooxy methyl-4-nitrobenzene hydrochloride, hydroxylamine, o-p-nitrophenylmethyl-, monohydrochloride | |
hydrogen O-[(4-nitrophenyl)methyl]hydroxylamine chloride |
Safety and Handling
RTECSNumber : NC4400050
TSCA : Yes