Learn More
O-Acetylsalicylic acid, 99%, Thermo Scientific Chemicals
$52.37 - $693.22
Chemical Identifiers
CAS | 50-78-2 |
---|---|
Molecular Formula | C9H8O4 |
Molecular Weight (g/mol) | 180.16 |
MDL Number | MFCD00002430 |
InChI Key | BSYNRYMUTXBXSQ-UHFFFAOYSA-N |
Synonym | aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin |
PubChem CID | 2244 |
ChEBI | CHEBI:15365 |
IUPAC Name | 2-acetyloxybenzoic acid |
SMILES | CC(=O)OC1=CC=CC=C1C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1248822
|
Thermo Scientific Chemicals
A1248822 |
100 g |
Each for $52.37
|
|
|||||
AAA1248836
|
Thermo Scientific Chemicals
A1248836 |
500 g |
Each for $199.12
|
|
|||||
AAA124880E
|
Thermo Scientific Chemicals
A124880E |
2500 g |
Each for $693.22
|
|
|||||
Description
O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.
Solubility
Soluble in 100% ethanol (80mg/ml), DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS), pH 7.2 (2.7mg/ml).
Notes
Store at room temperature. Ensure good ventilation.
Chemical Identifiers
50-78-2 | |
180.16 | |
BSYNRYMUTXBXSQ-UHFFFAOYSA-N | |
2244 | |
2-acetyloxybenzoic acid |
C9H8O4 | |
MFCD00002430 | |
aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin | |
CHEBI:15365 | |
CC(=O)OC1=CC=CC=C1C(O)=O |
Specifications
50-78-2 | |
1.4 | |
C9H8O4 | |
100 g | |
14,851 | |
Soluble in 100% ethanol (80mg/ml),DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS),pH 7.2 (2.7mg/ml). | |
CC(=O)OC1=CC=CC=C1C(O)=O | |
180.16 | |
CHEBI:15365 | |
99% |
133°C to 140°C | |
250°C (482°F) | |
MFCD00002430 | |
779271 | |
aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin | |
BSYNRYMUTXBXSQ-UHFFFAOYSA-N | |
2-acetyloxybenzoic acid | |
2244 | |
180.15 | |
O-Acetylsalicylic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302-H500
EINECSNumber : 200-064-1
RTECSNumber : VO0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only