missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Anisaldehyde, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 149220050

Catalog No. AC149220050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

o-Anisaldehyde
135-02-4
100.0
117°C
97.5% min. (GC)
C8H8O2
CH3OC6H4CHO
5g
10, 5866
Solubility in water: insoluble. Other solubilities: soluble in alcohol
COC1=CC=CC=C1C=O
136.15
136.15
98%
97.5
238.0°C
Authentic
Glass bottle
1.5608
MFCD00003308
08, IV, 180
o-anisaldehyde, benzaldehyde, 2-methoxy, 2-anisaldehyde, o-methoxybenzaldehyde, 6-methoxybenzaldehyde, salicylaldehyde methyl ether, 2-methoxybenzenecarboxaldehyde, benzaldehyde, o-methoxy, 2-methoxyphenylformaldehyde, formylanisole, o
PKZJLOCLABXVMC-UHFFFAOYSA-N
2-methoxybenzaldehyde
8658
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 205-171-7

RTECSNumber : BZ2610000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only