missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Phenetidine 98.0+%, TCI America™

Supplier:  TCI America P0089500G

 View more versions of this product

Catalog No. P0089500G


Explore available promotions

Description

Description

Specifications

Specifications

o-Phenetidine
-4°C
232°C
MFCD00007689
2311
ULHFFAFDSSHFDA-UHFFFAOYSA-N
2-ethoxyaniline
7203
≥98.0% (GC)
94-70-2
Yellow
C8H11NO
500 g
o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
CCOC1=CC=CC=C1N
137.182
137.18
Liquid

Chemical Identifiers

94-70-2
137.182
ULHFFAFDSSHFDA-UHFFFAOYSA-N
7203
CCOC1=CC=CC=C1N
C8H11NO
MFCD00007689
o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
2-ethoxyaniline
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0682&(3)-2773

RTECSNumber : SI6465300

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions