missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-Phosphorylethanolamine, 98%, Thermo Scientific Chemicals
$162.73 - $162.73
Chemical Identifiers
CAS | 1071-23-4 |
---|---|
Molecular Formula | C2H8NO4P |
Molecular Weight (g/mol) | 141.06 |
MDL Number | MFCD00008178 |
InChI Key | SUHOOTKUPISOBE-UHFFFAOYSA-N |
Synonym | o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate |
PubChem CID | 1015 |
ChEBI | CHEBI:17553 |
IUPAC Name | 2-aminoethyl dihydrogen phosphate |
SMILES | C(COP(=O)(O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC367420250
|
Thermo Scientific Chemicals
367420250 |
25 g | Glass Bottle |
Each for $162.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1071-23-4 | |
141.06 | |
SUHOOTKUPISOBE-UHFFFAOYSA-N | |
1015 | |
2-aminoethyl dihydrogen phosphate |
C2H8NO4P | |
MFCD00008178 | |
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate | |
CHEBI:17553 | |
C(COP(=O)(O)O)N |
Specifications
1071-23-4 | |
97.5 % min. (Titration with HClO4) | |
C2H8NO4P | |
MFCD00008178 | |
04,IV,1415 | |
Solubility in water: soluble. | |
C(COP(=O)(O)O)N | |
141.06 | |
CHEBI:17553 | |
98% |
241.0°C to 243.0°C | |
Glass Bottle | |
H2NCH2CH2OP(O)(OH)2 | |
25 g | |
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate | |
SUHOOTKUPISOBE-UHFFFAOYSA-N | |
2-aminoethyl dihydrogen phosphate | |
1015 | |
141.06 | |
O-Phosphorylethanolamine, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS Signal Word: Danger
EINECSNumber : 213-988-5
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only