missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 98.0+%, TCI America™
Supplier: TCI America B316825G
Specifications
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine | |
136°C | |
C22H24NO5 | |
25 g | |
REITVGIIZHFVGU-IBGZPJMESA-M | |
(2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate | |
2724633 | |
≥98.0% (HPLC,T) |
71989-33-8 | |
White | |
MFCD00037127 | |
fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh | |
CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O | |
382.44 | |
383.44 | |
Crystalline Powder |
Chemical Identifiers
71989-33-8 | |
382.44 | |
REITVGIIZHFVGU-IBGZPJMESA-M | |
2724633 | |
CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
C22H24NO5 | |
MFCD00037127 | |
fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh | |
(2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate |
Safety and Handling
TSCA : No