missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-tert-Butyl-N-Fmoc-L-beta-homoserine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H52065MD

 View more versions of this product

Catalog No. AAH52065MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

O-tert-Butyl-N-Fmoc-L-beta-homoserine
96°C to 98°C
MFCD01862865
8014825
SKYFRYZPXHNPOK-OAHLLOKOSA-N
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]butanoic acid
2761544
95%
203854-51-7
C23H27NO5
250 mg
fmoc-beta-hoser tbu-oh, fmoc-o-t-butyl-l-beta-homoserine, fmoc-l-beta-homoserine otbu, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy butanoic acid, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6740, fmoc-hoser tbu-oh, fmoc-??-hoser tbu-oh, fmoc-beta-homoser tbu-oh, fmoc-i 2-hoser tbu-oh
CC(C)(C)OCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
397.471
397.47

Chemical Identifiers

203854-51-7
397.471
SKYFRYZPXHNPOK-OAHLLOKOSA-N
2761544
CC(C)(C)OCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C23H27NO5
MFCD01862865
fmoc-beta-hoser tbu-oh, fmoc-o-t-butyl-l-beta-homoserine, fmoc-l-beta-homoserine otbu, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy butanoic acid, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6740, fmoc-hoser tbu-oh, fmoc-??-hoser tbu-oh, fmoc-beta-homoser tbu-oh, fmoc-i 2-hoser tbu-oh
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]butanoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only