Learn More
o-Toluic acid, 98+%
CAS: 118-90-1 | C8H8O2 | 136.15 g/mol
$66.92 - $167.53
Chemical Identifiers
| CAS | 118-90-1 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00002477 |
| InChI Key | ZWLPBLYKEWSWPD-UHFFFAOYSA-N |
| Synonym | o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate |
| PubChem CID | 8373 |
| ChEBI | CHEBI:36632 |
| IUPAC Name | 2-methylbenzoic acid |
| SMILES | CC1=CC=CC=C1C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1385630
|
Thermo Scientific Chemicals
A1385630 |
250 g |
N/A
|
|
|||||
|
AAA138560B
|
Thermo Scientific Chemicals
A138560B |
1000 g |
N/A
|
|
|||||
Description
o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.
Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases.
Chemical Identifiers
| 118-90-1 | |
| 136.15 | |
| ZWLPBLYKEWSWPD-UHFFFAOYSA-N | |
| 8373 | |
| 2-methylbenzoic acid |
| C8H8O2 | |
| MFCD00002477 | |
| o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate | |
| CHEBI:36632 | |
| CC1=CC=CC=C1C(=O)O |
Specifications
| 118-90-1 | |
| 1.06 | |
| 148°C (298°F) | |
| C8H8O2 | |
| 250 g | |
| 14,9535 | |
| Soluble in alcohol and chloroform. Slightly soluble in water. | |
| CC1=CC=CC=C1C(=O)O | |
| 136.15 | |
| CHEBI:36632 | |
| ≥98% |
| 103°C to 106°C | |
| 258°C to 259°C | |
| Aromatic | |
| MFCD00002477 | |
| 1072103 | |
| o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate | |
| ZWLPBLYKEWSWPD-UHFFFAOYSA-N | |
| 2-methylbenzoic acid | |
| 8373 | |
| 136.15 | |
| o-Toluic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 204-284-9
RTECSNumber : XU1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only