missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Toluic hydrazide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0066806

 View more versions of this product

Catalog No. AAL0066806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

o-Toluic hydrazide
122°C to 124°C
MFCD00014756
1862809
KFXLXEQCRFGDRU-UHFFFAOYSA-N
2-methylbenzohydrazide
82110
≥98%
7658-80-2
C8H10N2O
5 g
o-toluic hydrazide, o-toluic acid hydrazide, o-toluohydrazide, 2-toluic hydrazide, 2-methyl-benzoic acid hydrazide, benzoylhydrazine, o-methyl, benzoic acid, 2-methyl-, hydrazide, 2-methylbenzenecarbohydrazide, methylbenzohydrazide, 2-methylbenzhydrazide
CC1=CC=CC=C1C(=O)NN
150.181
150.18

Chemical Identifiers

7658-80-2
150.181
KFXLXEQCRFGDRU-UHFFFAOYSA-N
82110
CC1=CC=CC=C1C(=O)NN
C8H10N2O
MFCD00014756
o-toluic hydrazide, o-toluic acid hydrazide, o-toluohydrazide, 2-toluic hydrazide, 2-methyl-benzoic acid hydrazide, benzoylhydrazine, o-methyl, benzoic acid, 2-methyl-, hydrazide, 2-methylbenzenecarbohydrazide, methylbenzohydrazide, 2-methylbenzhydrazide
2-methylbenzohydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-623-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only