missing translation for 'onlineSavingsMsg'
Learn More

o-Toluidine, 99%

CAS: 95-53-4 | C7H9N | 107.15 g/mol

$89.41 - $288.58

Chemical Identifiers

CAS 95-53-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.15
MDL Number MFCD00007730
InChI Key RNVCVTLRINQCPJ-UHFFFAOYSA-N
Synonym o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
PubChem CID 7242
ChEBI CHEBI:66892
IUPAC Name 2-methylaniline
SMILES CC1=CC=CC=C1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC139085000
SDS View Documents
Thermo Scientific Chemicals
139085000
500 mL Glass bottle N/A
Only null left
 
AC139080025
SDS View Documents
Thermo Scientific Chemicals
139080025
2.5 L Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-53-4
107.15
RNVCVTLRINQCPJ-UHFFFAOYSA-N
7242
2-methylaniline
C7H9N
MFCD00007730
o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
CHEBI:66892
CC1=CC=CC=C1N

Specifications

95-53-4
Colorless to Yellow
199.0°C to 200.0°C
Authentic
Glass bottle
1.5710 to 1.5730
MFCD00007730
12,772
15,9696
Solubility in water: 1.5g/100mL (25°C). Other solubilities: solible in diluted acids,soluble in alcohol and ether,miscible with ethanol
CC1=CC=CC=C1N
107.15
CHEBI:66892
99%
o-Toluidine, 99%
-23.0°C
1.0000g/mL
85°C
98.5% min. (GC)
C7H9N
CH3C6H4NH2
500 mL
1
o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
RNVCVTLRINQCPJ-UHFFFAOYSA-N
2-methylaniline
7242
107.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Toxic if inhaled.
Suspected of causing genetic defects if inhaled.
May cause cancer by inhalat

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau

GHS Signal Word: Danger

EINECSNumber : 202-429-0

RTECSNumber : XU2975000

TSCA : TSCA

Recommended Storage : This substance darkens on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only