Learn More
o-Tolylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
$240.02 - $240.02
Chemical Identifiers
| CAS | 932-31-0 |
|---|---|
| Molecular Formula | C7H7BrMg |
| Molecular Weight (g/mol) | 195.34 |
| MDL Number | MFCD00010350 |
| InChI Key | DVXDIGKNJYSMFM-UHFFFAOYSA-M |
| Synonym | o-tolylmagnesium bromide, grignard reagent, bromo 2-methylphenyl magnesium, bromo 2-methylphenyl magnesium, o-methylphenylmagnesium bromide, 2-methylphenylmagnesium bromide, magnesium,bromo 2-methylphenyl, o-tolyl magnesium bromide, o-tolyl magnesium bromide, o-tolylmagnesiumbromid, o-tolylmagnesiumbromid |
| PubChem CID | 101919 |
| IUPAC Name | magnesium;methylbenzene;bromide |
| SMILES | CC1=CC=CC=C1[Mg]Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC427471000
|
Thermo Scientific Chemicals
427471000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $240.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 932-31-0 | |
| 195.34 | |
| DVXDIGKNJYSMFM-UHFFFAOYSA-M | |
| 101919 | |
| CC1=CC=CC=C1[Mg]Br |
| C7H7BrMg | |
| MFCD00010350 | |
| o-tolylmagnesium bromide, grignard reagent, bromo 2-methylphenyl magnesium, bromo 2-methylphenyl magnesium, o-methylphenylmagnesium bromide, 2-methylphenylmagnesium bromide, magnesium,bromo 2-methylphenyl, o-tolyl magnesium bromide, o-tolyl magnesium bromide, o-tolylmagnesiumbromid, o-tolylmagnesiumbromid | |
| magnesium;methylbenzene;bromide |
Specifications
| 932-31-0,60-29-7 | |
| 1.0130g/mL | |
| AcroSeal™ Glass Bottle | |
| CH3C6H4MgBr | |
| 100 mL | |
| o-tolylmagnesium bromide, grignard reagent, bromo 2-methylphenyl magnesium, bromo 2-methylphenyl magnesium, o-methylphenylmagnesium bromide, 2-methylphenylmagnesium bromide, magnesium,bromo 2-methylphenyl, o-tolyl magnesium bromide, o-tolyl magnesium bromide, o-tolylmagnesiumbromid, o-tolylmagnesiumbromid | |
| CC1=CC=CC=C1[Mg]Br | |
| 195.34 | |
| 195.34 | |
| o-Tolylmagnesium bromide, 2M solution in diethyl ether |
| Brown to Gray | |
| −40°C | |
| C7H7BrMg | |
| MFCD00010350 | |
| 1.013 | |
| DVXDIGKNJYSMFM-UHFFFAOYSA-M | |
| magnesium;methylbenzene;bromide | |
| 101919 | |
| Solution |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 213-250-2
RUO – Research Use Only