missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Vanillin, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 153071000

 View more versions of this product

Catalog No. AC153071000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

o-Vanillin
148-53-8
Brown to Yellow
>112°C
98.5% min. (GC)
C8H8O3
100 g
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
JJVNINGBHGBWJH-UHFFFAOYSA-N
2-hydroxy-3-methoxybenzaldehyde
8991
152.15
Low Melting Solid
99%
40.0°C to 42.0°C
265.0°C to 266.0°C
Authentic
Glass bottle
MFCD00003322
08, IV, 1747
Solubility in water: slightly soluble. Other solubilities: very soluble in alcohol,ether,benzene,carbon,disulfide
COC1=CC=CC(C=O)=C1O
152.15
CHEBI:78339
99%

Chemical Identifiers

148-53-8
152.15
JJVNINGBHGBWJH-UHFFFAOYSA-N
8991
2-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00003322
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
CHEBI:78339
COC1=CC=CC(C=O)=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 205-715-3

RTECSNumber : CU6530000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only