Learn More
o-Vanillin, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 153075000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
o-Vanillin | |
148-53-8 | |
Brown to Yellow | |
>112°C | |
98.5% min. (GC) | |
C8H8O3 | |
500 g | |
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde | |
JJVNINGBHGBWJH-UHFFFAOYSA-N | |
2-hydroxy-3-methoxybenzaldehyde | |
8991 | |
152.15 | |
Low Melting Solid |
99% | |
40.0°C to 42.0°C | |
265.0°C to 266.0°C | |
Authentic | |
Glass bottle | |
MFCD00003322 | |
08, IV, 1747 | |
Solubility in water: slightly soluble. Other solubilities: very soluble in alcohol,ether,benzene,carbon,disulfide | |
COC1=CC=CC(C=O)=C1O | |
152.15 | |
CHEBI:78339 | |
99% |
Chemical Identifiers
148-53-8 | |
152.15 | |
JJVNINGBHGBWJH-UHFFFAOYSA-N | |
8991 | |
2-hydroxy-3-methoxybenzaldehyde |
C8H8O3 | |
MFCD00003322 | |
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde | |
CHEBI:78339 | |
COC1=CC=CC(C=O)=C1O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 205-715-3
RTECSNumber : CU6530000
TSCA : TSCA
RUO – Research Use Only