missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Xylene (Certified), Fisher Chemical

Supplier:  Thermo Fisher Scientific O50814

Catalog No. O5081-4

Explore available promotions

Description

Description

Specifications

Specifications

o-Xylene
95-47-6,100-41-4
0.878g/cm³
4 L
Glass Bottle
C8H10
MFCD00008519
CTQNGGLPUBDAKN-UHFFFAOYSA-N
1,2-xylene
7237
106.16
-25°C
Colorless
143°C
Pass Test
Liquid
C6H4(CH3)2
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CC1=CC=CC=C1C
106.17
CHEBI:28063
Certified

Chemical Identifiers

95-47-6
106.17
CTQNGGLPUBDAKN-UHFFFAOYSA-N
7237
1,2-xylene
C8H10
MFCD00008519
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CHEBI:28063
CC1=CC=CC=C1C
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

SDS Documents

Documents

Accessories
Promotions

Promotions