Learn More
o-Xylene, Spectrophotometric Grade, 96% min, Thermo Scientific Chemicals
$202.73 - $549.34
Chemical Identifiers
CAS | 95-47-6 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008519 |
InChI Key | CTQNGGLPUBDAKN-UHFFFAOYSA-N |
Synonym | o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes |
PubChem CID | 7237 |
ChEBI | CHEBI:28063 |
IUPAC Name | 1,2-xylene |
SMILES | CC1=CC=CC=C1C |
Description
o-Xylene is used as a solvent for paints and adhesives, and to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-47-6 | |
106.17 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
7237 | |
1,2-xylene |
C8H10 | |
MFCD00008519 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CHEBI:28063 | |
CC1=CC=CC=C1C |
Specifications
-25°C | |
Colorless | |
143°C to 145°C | |
32°C (89°F) | |
1.505 | |
(CH3)2C6H4 | |
UN1307 | |
14,10081 | |
Sparingly soluble in water. (0.2g/L) | |
CC1=CC=CC=C1C | |
106.17 | |
CHEBI:28063 | |
≤96% | |
o-Xylene |
95-47-6 | |
0.877 g/mL | |
1 L | |
Liquid | |
C8H10 | |
MFCD00008519 | |
1815558 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
7237 | |
106.17 | |
Spectrophotometric |
Safety and Handling
GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
H226-H304-H312+H332-H315-H319-H335-H336-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
EINECSNumber : 202-422-2
RTECSNumber : ZE2450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only